Assay Details
| Gene Symbol: Ttbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107766.1 | NP_001101236.1 | 12 - 13 | 2083 | 57 | |
| XM_006234833.2 | XP_006234895.1 | 14 - 15 | 2516 | 57 | ||
| XM_006234834.2 | XP_006234896.1 | 14 - 15 | 2159 | 57 | ||
| XM_006234835.2 | XP_006234897.1 | 13 - 14 | 2442 | 57 | ||
| XM_006234836.2 | XP_006234898.1 | 12 - 13 | 2294 | 57 | ||
| XM_006234837.2 | XP_006234899.1 | 14 - 15 | 2162 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttbk2
Genomic Map