Assay Details
| Gene Symbol: Ppan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011980.1 | NP_001011980.1 | 7 - 8 | 786 | 66 | |
| GenBank mRNA | BC089842.1 | - | 7 - 8 | 786 | 7366262 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Ppan
Genomic Map