Assay Details
| Gene Symbol: Mrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106123.2 | NP_001099593.2 | 8 - 9 | 1516 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P18738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45857 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18738 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18738 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P18738 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P635138 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18738 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37177 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10796485 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796499 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796487 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796479 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796501 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796488 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796496 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796504 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796486 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796495 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796502 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796482 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796491 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796480 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796478 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796500 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796498 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796481 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796489 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796503 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796497 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796505 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796483 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796490 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796493 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796484 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796492 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796494 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796477 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10796476 | - | - |
| Rat Genome 230 2.0 Array | 1392648_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392648_at | - | - |
| Affymetrix HT Rat Focus Array | 1392648_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392648_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700007233.rn.1 | TC1700001055.rn.1 | RTA TC |
| Rat Expression Set 230 | 1392648_PM_at | - | - |
| Rat Expression Set 230 | 1392648_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796476 | - | - |
| Rat Genome U34 Set | rc_AA957139_at | - | - |
| Rat Genome U34 Set | rc_AI234748_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21426003 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374846 | - | - |

Mrc1
Genomic Map