Assay Details
Gene Symbol: Tssk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271325.1 | NP_001258254.1 | 3 - 4 | 955 | 91 | |
NR_073162.1 | 3 - 4 | 1011 | 91 | |||
GenBank mRNA | BC079202.1 | - | 3 - 4 | 1011 | 7131733 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
