Assay Details
| Gene Symbol: Cmtm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106034.2 | NP_001099504.2 | 2 - 3 | 615 | 63 | |
| GenBank mRNA | CX569344.1 | - | 1 - 2 | 131 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cmtm5
Genomic Map