Assay Details
| Gene Symbol: Rnf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108868.1 | NP_001102338.2 | 11 - 12 | 2216 | 73 | |
| XM_006252000.2 | XP_006252062.1 | 11 - 12 | 2914 | 73 | ||
| GenBank mRNA | BC169074.1 | - | 3 - 4 | 454 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf31
Genomic Map