Assay Details
| Gene Symbol: RGD1306227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014037.1 | NP_001014059.1 | 11 - 12 | 1867 | 123 | |
| GenBank mRNA | BC078991.1 | - | 11 - 12 | 1867 | 7112582 | 123 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1306227
Genomic Map