Assay Details
| Gene Symbol: Cdk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100506.1 | NP_001093976.1 | 4 - 5 | 397 | 69 | |
| XM_006249750.2 | XP_006249812.1 | 5 - 6 | 484 | 69 | ||
| XM_006249751.2 | XP_006249813.1 | 5 - 6 | 545 | 69 | ||
| XM_006249752.2 | XP_006249814.1 | 5 - 6 | 1005 | 69 | ||
| XM_006249753.2 | XP_006249815.1 | 5 - 6 | 518 | 69 | ||
| XM_008769441.1 | XP_008767663.1 | 5 - 6 | 823 | 69 | ||
| GenBank mRNA | AB005541.1 | - | 5 - 6 | 477 | 69 | |
| BC082045.1 | - | 4 - 5 | 397 | 7131722 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk18
Genomic Map