Assay Details
| Gene Symbol: Dars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034143.1 | NP_001029315.1 | 16 - 17 | 1851 | 131 | |
| GenBank mRNA | BC105774.1 | - | 16 - 17 | 1851 | 7930977 | 131 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1017913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164928 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1017913 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1017913 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1017913 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1017913 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P313904 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38174 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74900 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104301 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96969 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10769199 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769193 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769188 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769194 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769189 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769198 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769203 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769201 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769190 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769191 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769196 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769200 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769202 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769195 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769187 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769192 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769197 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10769186 | - | - |
| Rat Genome 230 2.0 Array | 1376135_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376135_at | - | - |
| Rat Genome 230 2.0 Array | 1398475_at | - | - |
| Rat Genome 230 2.0 Array | 1398475_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1398475_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376135_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1398475_at | - | - |
| Affymetrix HT Rat Focus Array | 1376135_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300012218.rn.1 | TC1300001747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300012219.rn.1 | TC1300001747.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376135_at | - | - |
| Rat Expression Set 230 | 1376135_PM_at | - | - |
| Rat Expression Set 230 | 1398475_at | - | - |
| Rat Expression Set 230 | 1398475_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769186 | - | - |
| Rat Genome U34 Set | rc_AA901017_at | - | - |
| Rat Genome U34 Set | rc_AI172076_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21195756 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373237 | - | - |

Dars2
Genomic Map