Assay Details
Gene Symbol: Zp3r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270844.1 | NP_001257773.1 | 6 - 7 | 807 | 87 | |
NM_001270845.1 | NP_001257774.1 | 6 - 7 | 729 | 87 | ||
NM_182815.2 | NP_877967.2 | 7 - 8 | 924 | 87 | ||
XM_006249713.2 | XP_006249775.1 | 7 - 8 | 1017 | 87 | ||
XM_008769436.1 | XP_008767658.1 | 6 - 7 | 939 | 87 | ||
XM_008769437.1 | XP_008767659.1 | 7 - 8 | 940 | 87 | ||
XM_008769438.1 | XP_008767660.1 | 7 - 8 | 923 | 87 | ||
XM_008769439.1 | XP_008767661.1 | 6 - 7 | 823 | 87 | ||
XM_008769440.1 | XP_008767662.1 | 7 - 8 | 1110 | 87 | ||
GenBank mRNA | AY278363.1 | - | 7 - 8 | 924 | 87 | |
AY278364.1 | - | 6 - 7 | 729 | 87 | ||
BC079287.1 | - | 6 - 7 | 786 | 7134311 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
