Assay Details
| Gene Symbol: Tex35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001055979.4 | XP_001055979.2 | 5 - 6 | 347 | 115 | |
| XM_006250091.2 | XP_006250153.1 | 5 - 6 | 461 | 115 | ||
| XM_006250092.2 | XP_006250154.1 | 5 - 6 | 461 | 115 | ||
| XM_008769693.1 | XP_008767915.1 | 5 - 6 | 461 | 115 | ||
| XM_008769694.1 | XP_008767916.1 | 5 - 6 | 461 | 115 | ||
| XM_008769695.1 | XP_008767917.1 | 4 - 5 | 404 | 115 | ||
| XM_008769696.1 | XP_008767918.1 | 5 - 6 | 461 | 115 | ||
| XM_008769697.1 | XP_008767919.1 | 5 - 6 | 347 | 115 | ||
More Information
Set Membership: |
|

Tex35
Genomic Map