Assay Details
| Gene Symbol: Sncaip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107379.1 | NP_001100849.1 | 7 - 8 | 1785 | 68 | |
| XM_006254719.2 | XP_006254781.1 | 7 - 8 | 1771 | 68 | ||
| XM_006254720.2 | XP_006254782.1 | 7 - 8 | 1522 | 68 | ||
| XM_006254721.2 | XP_006254783.1 | 7 - 8 | 1616 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sncaip
Genomic Map