Assay Details
| Gene Symbol: Prr16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108432.1 | NP_001101902.1 | 1 - 2 | 356 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P026923 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P026928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137008 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026928 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026923 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026923 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026928 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P026923 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P026928 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P229555 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99153 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15829 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88328 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89046 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31573 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68291 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10801686 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10801685 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10801684 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10801683 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1800003676.rn.1 | TC1800000500.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800003679.rn.1 | TC1800000500.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800003677.rn.1 | TC1800000500.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800003678.rn.1 | TC1800000500.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10801683 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21380354 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364166 | - | - |

Prr16
Genomic Map