Assay Details
| Gene Symbol: Srek1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008373.1 | NP_001008374.1 | 109 | 61 | ||
| GenBank mRNA | AY236996.1 | - | 150 | 61 | ||
| BC086553.1 | - | 109 | 7303262 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AY236996.1,BC086553.1,NM_001008373.1 transcripts.
This assay is in the 5'UTR region of the AY236996.1,BC086553.1,NM_001008373.1 transcripts.
Set Membership: |
|

Srek1ip1
Genomic Map