Assay Details
| Gene Symbol: Sgtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181629.3 | NP_853660.1 | 3 - 4 | 300 | 64 | |
| XM_006231882.2 | XP_006231944.1 | 3 - 4 | 466 | 64 | ||
| XM_006231883.2 | XP_006231945.1 | 3 - 4 | 441 | 64 | ||
| GenBank mRNA | AF368280.1 | - | 3 - 4 | 301 | 64 | |
| FQ212077.1 | - | 3 - 4 | 468 | 64 | ||
| FQ212215.1 | - | 3 - 4 | 443 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sgtb
Genomic Map