Assay Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191958.1 | NP_001178887.1 | 10 - 11 | 3767 | 68 | |
| XM_006231783.2 | XP_006231845.1 | 9 - 10 | 3677 | 68 | ||
| XM_006231784.2 | XP_006231846.1 | 11 - 12 | 3850 | 68 | ||
| XM_006231785.2 | XP_006231847.1 | 10 - 11 | 3736 | 68 | ||
| XM_008760640.1 | XP_008758862.1 | 10 - 11 | 4114 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfyve16
Genomic Map