Assay Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014253.2 | NP_001014275.2 | 1 - 2 | 191 | 94 | |
| GenBank mRNA | AY995234.1 | - | 1 - 2 | 191 | 94 | |
| BC086953.1 | - | 1 - 2 | 85 | 7321890 | 93 | |
| FQ211813.1 | - | 209 | 94 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC086953.1 transcripts.
Set Membership: |
|

Selt
Genomic Map