Assay Details
| Gene Symbol: Krt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153476.1 | NP_703206.1 | 2 - 3 | 718 | 114 | |
| GenBank mRNA | AY128946.1 | - | 2 - 3 | 718 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt9
Genomic Map