Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011894.1 | NP_001011894.1 | 3 - 4 | 340 | 103 | |
| GenBank mRNA | BC089889.1 | - | 3 - 4 | 340 | 7374359 | 103 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psme3
Genomic Map