Assay Details
| Gene Symbol: Fam20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012237.1 | NP_001012237.1 | 10 - 11 | 1358 | 63 | |
| XM_006247610.2 | XP_006247672.1 | 10 - 11 | 1460 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P516194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130172 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P516194 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P516194 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P516194 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P516194 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3446 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84382 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10748547 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748546 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748554 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748553 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748552 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748551 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748548 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748555 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748549 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748545 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748550 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10748544 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000036529.rn.1 | TC1000004032.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10748544 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21923017 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357332 | - | - |

Fam20a
Genomic Map