Assay Details
| Gene Symbol: P2ry10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177682.1 | NP_001171153.1 | 1 - 2 | 234 | 102 | |
| GenBank mRNA | BC168967.1 | - | 1 - 2 | 234 | 7381748 | 102 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC168967.1,NM_001177682.1 transcripts.
Set Membership: |
|

P2ry10
Genomic Map