Assay Details
| Gene Symbol: Robo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768537.1 | XP_008766759.1 | 8 - 9 | 2073 | 67 | |
| XM_008768538.1 | XP_008766760.1 | 8 - 9 | 2072 | 67 | ||
| XM_008768539.1 | XP_008766761.1 | 8 - 9 | 2069 | 67 | ||
| GenBank mRNA | AF182037.1 | - | 9 - 10 | 1910 | 67 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
metanephros development
ureteric bud development
homophilic cell adhesion via plasma membrane adhesion molecules
axon guidance
brain development
axon midline choice point recognition
spinal cord development
olfactory bulb interneuron development
retinal ganglion cell axon guidance
cellular response to hormone stimulus
positive regulation of axonogenesis
negative regulation of negative chemotaxis
apoptotic process involved in luteolysis
axon guidance receptor activity
identical protein binding
metanephros development
ureteric bud development
homophilic cell adhesion via plasma membrane adhesion molecules
axon guidance
brain development
axon midline choice point recognition
spinal cord development
olfactory bulb interneuron development
retinal ganglion cell axon guidance
cellular response to hormone stimulus
positive regulation of axonogenesis
negative regulation of negative chemotaxis
apoptotic process involved in luteolysis
axon guidance receptor activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P900672 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P041927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77719 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P900672 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P041927 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P041927 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P900672 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P041927 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P900672 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P900672 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P501942 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P584710 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700485984 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15662 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45190 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18382 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71982 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13887 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102328 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66602 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70086 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18896 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86479 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67044 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96120 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4920 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35557 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80927 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63095 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27215 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4721 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60887 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31172 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36885 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90872 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700485984 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10752703 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752712 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752711 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752720 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752718 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752707 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752710 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752709 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752721 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752716 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752714 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752719 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752708 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752705 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752723 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752713 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752706 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752717 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10752695 | - | - |
| Rat Genome 230 2.0 Array | 1382632_at | - | - |
| Rat Genome 230 2.0 Array | 1382632_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379693_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379750_at | - | - |
| Rat Genome 230 2.0 Array | 1379750_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379693_at | - | - |
| Affymetrix HT Rat Focus Array | 1382632_at | - | - |
| Affymetrix HT Rat Focus Array | 1379750_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379693_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379750_at | - | - |
| Affymetrix HT Rat Focus Array | 1382632_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379693_at | - | - |
| Rat Expression Set 230 | 1382632_at | - | - |
| Rat Expression Set 230 | 1382632_PM_at | - | - |
| Rat Expression Set 230 | 1379693_PM_at | - | - |
| Rat Expression Set 230 | 1379750_at | - | - |
| Rat Expression Set 230 | 1379750_PM_at | - | - |
| Rat Expression Set 230 | 1379693_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752695 | - | - |
| Rat Genome U34 Set | rc_AI639443_at | - | - |
| Rat Genome U34 Set | rc_AI228714_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20989151 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353494 | - | - |

Robo2
Genomic Map