Assay Details
| Gene Symbol: Pgam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025272.1 | NP_001020443.1 | 4 - 5 | 614 | 82 | |
| GenBank mRNA | BC092607.1 | - | 4 - 5 | 614 | 7309810 | 82 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pgam5
Genomic Map