Assay Details
| Gene Symbol: Myo18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249577.2 | XP_006249639.1 | 22 - 23 | 4355 | 58 | |
| XM_006249578.2 | XP_006249640.1 | 21 - 22 | 4144 | 58 | ||
More Information
Set Membership: |
|

Myo18b
Genomic Map