Assay Details
| Gene Symbol: Tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004204.2 | NP_001004204.1 | 5 - 6 | 715 | 130 | |
| XM_006249230.2 | XP_006249292.1 | 2 - 3 | 417 | 130 | ||
| XM_006249231.2 | XP_006249293.1 | 2 - 3 | 338 | 130 | ||
| GenBank mRNA | BC079161.1 | - | 2 - 3 | 389 | 7129411 | 130 |
| BC090318.1 | - | 5 - 6 | 715 | 7113430 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem248
Genomic Map