Assay Details
| Gene Symbol: Klrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001072378.4 | XP_001072378.2 | 1 - 2 | 1299 | 78 | |
| XM_006236344.2 | XP_006236406.1 | 1 - 2 | 1300 | 78 | ||
| XM_006236345.2 | XP_006236407.1 | 1 - 2 | 1300 | 78 | ||
More Information
Set Membership: |
|

Klrg2
Genomic Map