Assay Details
| Gene Symbol: Pdgfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031524.1 | NP_113712.1 | 4 - 5 | 854 | 65 | |
| GenBank mRNA | L40991.1 | - | 1 - 2 | 216 | 65 | |
| Z14117.1 | - | 4 - 5 | 435 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfb
Genomic Map