Assay Details
| Gene Symbol: Myf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106783.1 | NP_001100253.1 | 1 - 2 | 734 | 100 | |
| XM_006241306.2 | XP_006241368.1 | 1 - 2 | 616 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myf5
Genomic Map