Assay Details
| Gene Symbol: Hcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008357.1 | NP_001008358.1 | 12 - 13 | 1673 | 123 | |
| GenBank mRNA | BC085951.1 | - | 12 - 13 | 1673 | 7132601 | 123 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P042930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32316 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042930 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042930 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P042930 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P998555 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74750 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83061 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 54097 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22729 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59410 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10901454 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901450 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901452 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901447 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901449 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901456 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901444 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901453 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901446 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901448 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901445 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901451 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901443 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901457 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901455 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10901442 | - | - |
| Rat Genome 230 2.0 Array | 1392089_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392089_at | - | - |
| Affymetrix HT Rat Focus Array | 1392089_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392089_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700018974.rn.1 | TC0700002399.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700018972.rn.1 | TC0700002399.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700018973.rn.1 | TC0700002399.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700018975.rn.1 | TC0700002399.rn.1 | RTA TC |
| Rat Expression Set 230 | 1392089_at | - | - |
| Rat Expression Set 230 | 1392089_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901442 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367190 | - | - |
| RatRef-12 Expression BeadChip | ILMN_67013 | - | - |

Hcfc2
Genomic Map