Assay Details
| Gene Symbol: Epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108088.1 | NP_001101558.1 | 3 - 4 | 295 | 125 | |
| XM_006241286.2 | XP_006241348.1 | 2 - 3 | 658 | 125 | ||
| GenBank mRNA | BC161903.1 | - | 2 - 3 | 501 | 8367209 | 125 |
More Information
Updated Mapping Information
Set Membership: |
|

Epyc
Genomic Map