Assay Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133313.2 | NP_579847.1 | 10 - 11 | 1328 | 70 | |
| GenBank mRNA | AJ301634.1 | - | 10 - 11 | 1437 | 70 | |
| BC062087.1 | - | 10 - 11 | 1328 | 5621521 | 70 | |
| FQ213088.1 | - | 5 - 6 | 560 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atxn10
Genomic Map