Assay Details
| Gene Symbol: Fam109b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006242125.2 | XP_006242187.1 | 1 - 2 | 804 | 166 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006242125.2 transcripts.
Set Membership: |
|

Fam109b
Genomic Map