Assay Details
| Gene Symbol: Fam109b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130511.1 | NP_001123983.1 | 1 - 2 | 239 | 155 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001130511.1 transcripts.
Set Membership: |
|

Fam109b
Genomic Map