Assay Details
| Gene Symbol: RGD1310852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025007.1 | NP_001020178.1 | 3 - 4 | 412 | 104 | |
| GenBank mRNA | BC091232.1 | - | 3 - 4 | 412 | 7322456 | 104 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1310852
Genomic Map