Assay Details
Gene Symbol: Slx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009292.1 | NP_001009292.1 | 3 - 4 | 700 | 123 | |
GenBank mRNA | BC087090.1 | - | 3 - 4 | 700 | 7190632 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
