Assay Details
| Gene Symbol: LOC361635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017463.1 | NP_001017463.1 | 28 - 29 | 2575 | 77 | |
| XM_006230122.2 | XP_006230184.1 | 29 - 30 | 2773 | 77 | ||
| GenBank mRNA | BC083805.1 | - | 28 - 29 | 2575 | 7109216 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P001922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86420 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P001922 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P001922 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P001922 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43734 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66559 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47399 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65840 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60104 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58884 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10710213 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710222 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710196 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710202 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710216 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710199 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710203 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710221 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710220 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710195 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710198 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710208 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710217 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710197 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710210 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710214 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710207 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710223 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710212 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710219 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710209 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710225 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710201 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710218 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710200 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710204 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710206 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710205 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710211 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710224 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10710194 | - | - |
| Rat Genome 230 2.0 Array | 1367514_at | - | - |
| Rat Genome 230 2.0 Array | 1367514_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367514_at | - | - |
| Affymetrix HT Rat Focus Array | 1367514_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100018729.rn.1 | TC0100002473.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367514_PM_at | - | - |
| Rat Expression Set 230 | 1367514_at | - | - |
| Rat Expression Set 230 | 1367514_at | - | - |
| Rat Expression Set 230 | 1367514_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710194 | - | - |
| Rat Genome U34 Set | rc_AI235508_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21607909 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367362 | - | - |

LOC361635
Genomic Map