Assay Details
| Gene Symbol: Klhl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229818.1 | XP_006229880.1 | 2 - 3 | 4967 | 90 | |
| XM_218953.6 | XP_218953.4 | 1 - 2 | 967 | 90 | ||
More Information
Set Membership: |
|

Klhl35
Genomic Map