Assay Details
| Gene Symbol: St5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107547.1 | NP_001101017.1 | 11 - 12 | 2543 | 63 | |
| XM_006229976.2 | XP_006230038.1 | 12 - 13 | 3965 | 63 | ||
| XM_006229977.2 | XP_006230039.1 | 12 - 13 | 2560 | 63 | ||
| XM_006229979.2 | XP_006230041.1 | 11 - 12 | 3060 | 63 | ||
| XM_008759737.1 | XP_008757959.1 | 10 - 11 | 3730 | 63 | ||
| XM_008759738.1 | XP_008757960.1 | 12 - 13 | 3847 | 63 | ||
| XM_008759739.1 | XP_008757961.1 | 13 - 14 | 3951 | 63 | ||
| XM_008759740.1 | XP_008757962.1 | 9 - 10 | 3514 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

St5
Genomic Map