Assay Details
| Gene Symbol: Poc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729781.3 | XP_002729827.2 | 4 - 5 | 529 | 77 | |
| XM_008765341.1 | XP_008763563.1 | 4 - 5 | 694 | 77 | ||
| XM_008765342.1 | XP_008763564.1 | 3 - 4 | 761 | 77 | ||
| GenBank mRNA | FQ231893.1 | - | 4 - 5 | 593 | 77 | |
| FQ233318.1 | - | 4 - 5 | 593 | 77 | ||
| FQ235303.1 | - | 4 - 5 | 593 | 77 | ||
More Information
Set Membership: |
|

Poc1b
Genomic Map