Assay Details
| Gene Symbol: Mrpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133539.1 | NP_598223.1 | 1 - 2 | 154 | 63 | |
| GenBank mRNA | BC058447.1 | - | 1 - 2 | 199 | 6919782 | 63 |
| FQ212468.1 | - | 1 - 2 | 260 | 63 | ||
| FQ229319.1 | - | 1 - 2 | 212 | 63 | ||
| U53512.1 | - | 1 - 2 | 154 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_133539.1,U53512.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrpl17
Genomic Map