Assay Details
| Gene Symbol: Art5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013039.1 | NP_001013057.1 | 4 - 5 | 890 | 119 | |
| XM_006229843.2 | XP_006229905.1 | 2 - 3 | 476 | 119 | ||
| XM_006229845.2 | XP_006229907.1 | 2 - 3 | 478 | 119 | ||
| XM_006229846.2 | XP_006229908.1 | 3 - 4 | 508 | 119 | ||
| XM_008759692.1 | XP_008757914.1 | 2 - 3 | 420 | 119 | ||
| XM_008759693.1 | XP_008757915.1 | 2 - 3 | 476 | 119 | ||
| GenBank mRNA | BC083915.1 | - | 4 - 5 | 890 | 7134556 | 119 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC083915.1, NM_001013039.1, XM_006229843.2, XM_006229845.2, XM_006229846.2, XM_008759692.1, XM_008759693.1 transcripts.
Set Membership: |
|

Art5
Genomic Map