Assay Details
| Gene Symbol: Ccdc89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134833.2 | NP_001128305.1 | 1 | 1088 | 77 | |
| GenBank mRNA | BC167063.1 | - | 1 | 1088 | 7453194 | 77 |
| Gene Symbol: LOC100911776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748922.3 | XP_003748970.1 | 1 | 2628 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc89
Genomic Map