Assay Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748988.3 | XP_003749036.1 | 2 - 3 | 544 | 95 | |
| XM_008759928.1 | XP_008758150.1 | 1 - 2 | 233 | 95 | ||
| XM_008759929.1 | XP_008758151.1 | 1 - 2 | 233 | 95 | ||
| XM_008759930.1 | XP_008758152.1 | 1 - 2 | 233 | 95 | ||
| GenBank mRNA | BC166576.1 | - | 1 - 2 | 98 | 7321704 | 95 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166576.1, XM_003748988.3, XM_008759928.1, XM_008759929.1, XM_008759930.1 transcripts.
Set Membership: |
|

Tmem219
Genomic Map