Assay Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748988.3 | XP_003749036.1 | 5 - 6 | 1065 | 76 | |
| XM_008759928.1 | XP_008758150.1 | 4 - 5 | 751 | 76 | ||
| XM_008759929.1 | XP_008758151.1 | 3 - 4 | 556 | 76 | ||
| XM_008759930.1 | XP_008758152.1 | 3 - 4 | 553 | 76 | ||
| GenBank mRNA | BC166576.1 | - | 4 - 5 | 619 | 7321704 | 76 |
| FQ216353.1 | - | 1 - 2 | 463 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem219
Genomic Map