Assay Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748988.3 | XP_003749036.1 | 6 - 7 | 1256 | 87 | |
| XM_008759928.1 | XP_008758150.1 | 5 - 6 | 942 | 87 | ||
| XM_008759929.1 | XP_008758151.1 | 4 - 5 | 747 | 87 | ||
| XM_008759930.1 | XP_008758152.1 | 4 - 5 | 744 | 87 | ||
| GenBank mRNA | FQ216353.1 | - | 2 - 3 | 654 | 87 | |
More Information
Set Membership: |
|

Tmem219
Genomic Map