Assay Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748988.3 | XP_003749036.1 | 8 - 9 | 1654 | 105 | |
| XM_008759928.1 | XP_008758150.1 | 7 - 8 | 1340 | 105 | ||
| XM_008759929.1 | XP_008758151.1 | 6 - 7 | 1145 | 105 | ||
| XM_008759930.1 | XP_008758152.1 | 6 - 7 | 1142 | 105 | ||
| GenBank mRNA | FQ216353.1 | - | 4 - 5 | 1052 | 105 | |
More Information
Set Membership: |
|

Tmem219
Genomic Map