Assay Details
| Gene Symbol: Foxp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108788.1 | NP_001102258.1 | 10 - 11 | 1454 | 61 | |
| XM_006244461.2 | XP_006244523.1 | 11 - 12 | 1525 | 61 | ||
| XM_006244462.2 | XP_006244524.1 | 10 - 11 | 1481 | 61 | ||
| XM_008766866.1 | XP_008765088.1 | 11 - 12 | 1485 | 61 | ||
| XM_008766867.1 | XP_008765089.1 | 11 - 12 | 1482 | 61 | ||
| XM_008766868.1 | XP_008765090.1 | 11 - 12 | 1464 | 61 | ||
| XM_008766869.1 | XP_008765091.1 | 11 - 12 | 1428 | 61 | ||
| XM_008766870.1 | XP_008765092.1 | 11 - 12 | 1425 | 61 | ||
| XM_008766871.1 | XP_008765093.1 | 11 - 12 | 1461 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Foxp4
Genomic Map