Assay Details
| Gene Symbol: Ptcra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750652.3 | XP_003750700.1 | 2 - 3 | 982 | 74 | |
| XM_008766898.1 | XP_008765120.1 | 2 - 3 | 629 | 74 | ||
| GenBank mRNA | AF182508.1 | - | 1 - 2 | 201 | 74 | |
More Information
Set Membership: |
|

Ptcra
Genomic Map