Assay Details
| Gene Symbol: Ttc30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127607.1 | NP_001121079.1 | 1 | 2308 | 132 | |
| GenBank mRNA | BC166739.1 | - | 1 | 2308 | 7937387 | 132 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ttc30b
Genomic Map