Assay Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077429.1 | NP_001070897.1 | 2 - 3 | 149 | 151 | |
| XM_006234377.2 | XP_006234439.1 | 2 - 3 | 231 | 151 | ||
| XM_006234378.2 | XP_006234440.1 | 3 - 4 | 252 | 151 | ||
| XM_006234379.2 | XP_006234441.1 | 2 - 3 | 279 | 151 | ||
| XM_006234380.2 | XP_006234442.1 | 2 - 3 | 351 | 151 | ||
| XM_006234381.2 | XP_006234443.1 | 2 - 3 | 444 | 151 | ||
| XM_006234382.2 | XP_006234444.1 | 3 - 4 | 372 | 151 | ||
| XM_006234385.2 | XP_006234447.1 | 2 - 3 | 233 | 151 | ||
| XM_008761909.1 | XP_008760131.1 | 2 - 3 | 1328 | 151 | ||
| XM_008761910.1 | XP_008760132.1 | 2 - 3 | 233 | 151 | ||
| GenBank mRNA | BC126100.1 | - | 2 - 3 | 149 | 7132654 | 151 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC126100.1, NM_001077429.1, XM_006234377.2, XM_006234378.2, XM_006234379.2, XM_006234380.2, XM_006234381.2, XM_006234382.2, XM_006234385.2, XM_008761909.1, XM_008761910.1 transcripts.
Set Membership: |
|

Lnp
Genomic Map